Anti-CXCL8 Rabbit Polyclonal Antibody

Supplier: Abnova
ABNOPAB19092EA 0 INR
ABNOPAB19092
Anti-CXCL8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of IL8.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1 ?g/ml

Type: Primary
Antigen: CXCL8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR