Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triton® X-100 (Polyethylene glycol tert-octylphenyl ether)

Supplier: VWR Chemicals


R: 22-41-51/53
S: 26-39-61
H: H302 H318 H411
P: P280 P273 P305+P351+P338 P309+P310
Danger
 
  Boiling Pt: Min. 200 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1,06 g/cm³ (20 °C)
Flash Pt: 251 °C (closed cup)
CAS Number: 9002-93-1
ADR: 9,III
Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 1 l MX2347.290
Glass bottle 2,5 l MX2347.320
Metal drum 25 l MX2347.460
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty