Would you like to log in again?
Accessories for glove boxes, HEPA filtered, Precise™ and weigh boxes, XPert™
Supplier: Labconco
Precise®
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Replacement inlet or outlet HEPA filter, 99,99% efficiency (Ø 0,3 micron particles), weight 3 kg 1 LBCP5225105
Remote wall-mount blower kit, incl. epoxy-coated steel bracket, mounting hardware, hose, hose clamps and power cord extension for moving the integral blower from the glove box to a wall up to five feet away 1 135-1083
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty