Would you like to log in again?
Retort clamp
Supplier: R-L ENTERPRISES LTD
Alloy die-cast, aluminium shaft
This clamp accepts articles up to 85 mm diameter. Actuated by nickel-plated brass pummels and blue plastic headed nickel-plated steel twin screw.
  • Fitted with an aluminium shaft
  • Four interlocking jaws are cork lined
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Spanning width Pk VWR Catalog Number Quantity
85 1 212-9503
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty