Paraformaldehyde, prills, Reagent Grade

Synonyms: Polyoxymethylene

J531-500GEA 3692.25 INR
J531-500G
Paraformaldehyde, prills, Reagent Grade
Paraformaldehyde
Formula: (CH₂O)n
Melting Pt: 123 °C
MDL Number: MFCD00133991
CAS Number: 30525-89-4
ADR: 4.1,III
Merck Index: 13,07096

Order Now

Specification Test Results

Formaldehyde Weight 91.0 - 93.0 %
Iron ≤0.0002 %
Residue on Ignition ≤ 0.01 %
Water ≤9.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR