Digital timer clock, one channel
609-0186EA 1577 INR
VWRI609-0186
Digital timer clock, one channel
  • Format: 12/24 hours
  • Counts down and up for 23 hours, 59 minutes, 59 seconds
  • Separate hour, minute, second, start/stop and reset keys
  • Memory function
Certifications: Complies with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, accredited by A2LA indicates traceability to standards provided by NIST.
Delivery information: Supplied with foldaway stand, magnetic attachment, 1,5 V LR44 battery and certificate of calibration.