VWR, Cryogenic Cylinder With Mobile Head, XRP
478-0596EA 382907 INR
VWRI478-0596
VWR, Cryogenic Cylinder With Mobile Head, XRP
Flexible solution for the storage and transfer of liquid nitrogen in laboratories. This container is particularly adapted for liquid nitrogen and argon applications.
  • Equipped with a pressure regulator and a relief valve for a maximum work pressure till 1,5 bar
  • Fitted with an NW 50 flange on which a TAP withdrawing head can be fixed
Made in France complying with the highest quality standards. All the units are certified Medical CE-marked and comply with the Medical Device Directive 93/42/CEE and Pressure Equipment Directive – PED 97/23/CE.
Ordering information: other capacities or horizontal versions can be provided upon request.