1,10-Phenanthroline monohydrate, BAKER ANALYZED® ACS, J.T.Baker®
|
Danger
|
Synonyms:
1,10-Phenanthroline hydrate
T170-02EA
13460
INR
T170-02
1,10-Phenanthroline monohydrate, BAKER ANALYZED® ACS, J.T.Baker®
1,10-Phenanthroline monohydrate
Formula:
C₁₂H₈N₂·1H₂O MW: 198,22 g/mol Melting Pt: 93…94 °C Density: 1,259 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149973 CAS Number: 5144-89-8 EINECS: 200-629-2 UN: 2811 ADR: 6.1,III Merck Index: 12,07356 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Suitability as Redox Indicator | Passes Test |
Suitability for Determination of Iron | Passes Test |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...