Potassium hexacyanoferrate(II) trihydrate 98.5-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Synonyms: Tetrapotassium hexacyanoferrate trihydratePotassiumhexacyanoferrate(II) trihydratePotassium ferrocyanide trihydrate

3114-01EA 30438 INR
3114-01
Potassium hexacyanoferrate(II) trihydrate 98.5-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Potassium hexacyanoferrate (II) trihydrate

Appearance: mixture of white to slightly yellow plates and powder

Formula: K₄Fe(CN)₆·3H₂O
MW: 422,39 g/mol
Melting Pt: 70 °C
Density: 1,889 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (K₄Fe(CN)₆·3H₂O) 98.5 - 102.0 %
Insoluble Matter ≤0.005 %
Chloride (Cl) ≤0.01 %
Sulfate (SO₄) Passes Test
Appearance (mixture of white to slightly yellow plates and powder)
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR