Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3%, BAKER ANALYZED® ACS, J.T.Baker®
|
Danger
|
Synonyms:
Ethylene glycol monomethyl ether
P784-08EA
27923.44
INR
P784-08
P784-07
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.3%, BAKER ANALYZED® ACS, J.T.Baker®
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula:
C₃H₈O₂ MW: 76,1 g/mol Boiling Pt: 124,6 °C (1013 hPa) Melting Pt: –85 °C Density: 0,964 g/cm³ (20 °C) Flash Pt: 39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002867 CAS Number: 109-86-4 EINECS: 203-713-7 UN: 1188 ADR: 3,III Merck Index: 12,06118 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (C₃H₈O₂) (by GC, corrected for water) | ≥ 99.3 % |
Color (APHA) | ≤ 10 |
Titrable Acid (meq/g) | ≤ 0.002 |
Water (H₂O)(by Karl Fischer titrn) | ≤ 0.1 % |
pH (80% in Water), For Information Only | |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...