Bromocresol green, BAKER ANALYZED® ACS, J.T.Baker®

Warning

Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

BAKRC946-01EA 25010.57 INR
BAKRC946-01
Bromocresol green, BAKER ANALYZED® ACS, J.T.Baker®
Bromocresol green

Lot analysis on label.

Formula: C₂₁H₁₄Br₄O₅S
MW: 698,02 g/mol
Melting Pt: 217…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://in.vwr.com/stibo/web/std.lang.all/13/36/2301336.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Clarity of Solution Passes Test
Visual Transition Interval - pHÖ5.4 (Blue) Passes Test
Visual Transition Interval - pHÖ3.8 (Yellow) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR