Would you like to log in again?
Racks for microtubes
Supplier: VWR Collection
PP
  • Use with 1,5/2,0 ml microcentrifuge tubes and 2 ml cryotubes
  • Suitable for freezer use down to −80 °C
  • Autoclavable
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
For tubes No. of holes Colour Pk VWR Catalog Number Quantity
- - - 5 VWRU30128-274
1,5/2,0 80 (5×16) Assorted 5 VWRU30128-282
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description Pk VWR Catalog Number Quantity
Hinged storage box for 80-place racks 5 VWRU30128-290
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty