Ammonium heptamolybdate tetrahydrate (81.0 - 83.0% MoO₃), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Danger

Synonyms: Ammonium molybdate(VI) tetrahydrateAmmonium molybdate tetrahydrateMolybdic acid ammonium salt tetrahydrateHexaammonium heptamolybdate tetrahydrate

0716-01EA 39921 INR
0716-01
Ammonium heptamolybdate tetrahydrate (81.0 - 83.0% MoO₃), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Ammonium heptamolybdate tetrahydrate
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
MW: 1235,86 g/mol
Density: 2,50 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (as MoO₃) 81.0 - 83.0 %
Insoluble Matter ≤ 0.005 %
Chloride (Cl) ≤ 0.002 %
Nitrate (NO₃) Passes Test
Arsenate, Phosphate and Silicate (as SiO₂) ≤ 0.001 %
ACS - Phosphate (PO₄) ≤ 5 ppm
Sulfate (SO₄) ≤ 0.02 %
Heavy Metals (as Pb) ≤ 0.001 %
Magnesium (Mg) ≤ 0.005 %
Potassium (K) ≤ 0.01 %
Sodium (Na) ≤ 0.010 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR