Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Examination gloves, nitrile, Microflex® Supreno® EC

Supplier: MICROFLEX
Supreno® EC
Nitrile, non sterile, powder-free, textured fingertips

Supreno® EC provides extended-cuff protection of the wrists and arms for high risk applications. Designed to maintain a comfortable fit while providing reliable hazard protection, these gloves are also adhesive-resistant. Tested for use with chemotherapy drugs.

  • Ambidextrous
  • Textured fingertips for enhanced grip
  • Extended beaded cuffs
  • Very low pinhole rate (AQL 0,65)

Meets or exceeds NFPA requirements
ASTM D 6319; AQL 0,65




 

ORDER

Expand All / Collapse All
Glove size Colour Length Packed Pk VWR Catalog Number Quantity
S Blue 295 mm 50/box 50 MICFSEC-375-S
M Blue 295 mm 50/box 50 MICFSEC-375-M
L Blue 295 mm 50/box 50 MICFSEC-375-L
XL Blue 295 mm 50/box 50 MICFSEC-375-XL
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Ambidextrous/hand-specific Ambidextrous
Disposable/reusable Disposable
Latex/latex-free Latex-free
Powdered/powder-free Powder-free
Finish Textured fingertips
Cuff style Beaded
Cuff thickness 0,11 mm
Finger thickness 0,20 mm
Palm thickness 0,14 mm
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty