Would you like to log in again?
Single-stranded DNA binding protein (SSB)
Supplier: Affymetrix
Source: E. coli strain containing an overproducing clone of E. coli SSB protein.
Single-stranded DNA binding protein binds with high affinity in a cooperative manner to single-stranded DNA and does not bind well to double-stranded DNA. It is involved in DNA replication and recombination in vivo. This protein has been used to visualise single-stranded DNA by electron microscopy. It has also been used in conjunction with RecA protein for carrying out site-directed mutagenesis and to select sequences from libraries of double-stranded DNA. SSB may also stimulate specific DNA polymerases used in DNA sequencing reactions and has been used to target restriction endonuclease digestions to specific sites in single-stranded DNA for subsequent mutagenesis. SSB has been shown to be effective in fluorescence polarisation assays and eliminates pausing when sequencing through strong secondary structure. More recently, SSB was used to help obtain longer read lengths in pyrosequencing for SNP analysis.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Single stranded binding protein 500 µG USBM70032Z500UG
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty