Would you like to log in again?
Gloves, PE
Supplier: VWR BRAND (P/L-PRODUCTS USA)
Polyethylene, non sterile, embossed finish
These single-use gloves with extra-long cuffs provide excellent resistance to chemicals and detergents. Ideal for hand and product protection in virtually any type of laboratory, food handling, industrial safety, veterinary, electronics, painting, printing, or photography environments.
  • Ambidextrous
  • Embossed finish for improved grip
  • Tough, waterproof construction
  • Soft, virgin polyethylene gloves are so pliable that you can handle even small laboratory apparatus with ease
  • Convenient pull-out dispenser
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Size Colour Thickness Packed Pk VWR Catalog Number Quantity
S Transparent 0,03 mm 100/box 100 VWRU32915-224
M Transparent 0,03 mm 100/box 100 VWRU32915-246
L Transparent 0,03 mm 100/box 100 VWRU32915-268
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Ambidextrous/hand-specific Ambidextrous
Disposable/Reusable Disposable
Cuff style Straight
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty