Cleanroom tapes, TVR series
Supplier: Texwipe
TEXWTVR3016GREA 0 INR
TEXWTVR3016GR TEXWTVR3016BL TEXWTVR3016YE TEXWTVR2024WH TEXWTVR1048BL TEXWTVR1048YE TEXWTVR0596WH TEXWTVR1048GR TEXWTVR2024RD TEXWTVR0596RD TEXWTVR1048RD TEXWTVR1048WH TEXWTVR2024GR TEXWTVR2024BL TEXWTVR0596GR TEXWTVR0596BL TEXWTVR2024YE TEXWTVR0596YE TEXWTVR3016RD TEXWTVR3016WH
Cleanroom tapes, TVR series
PVC tape with synthetic rubber adhesive.
  • Latex-free
  • Ideal for cleanroom construction applications
  • Quick tack with abrasion resistance
  • Packaged in reusable press-and-seal cleanroom bags
Length: 32,9 m
Different colours available.