Horizontal electrophoresis systems, Mini-Fast Sub
Supplier: CBS Scientific
CBSCMGU-303EA 0 INR
CBSCMGU-303
Horizontal electrophoresis systems, Mini-Fast Sub
Mini-Fast Sub run times are less than an hour. The UV transparent base allows sample visualisation simply by transferring the unit to a transilluminator. The uniplanar design has a buffer chamber only slightly longer than the gel itself, minimising the amount of buffer required - 50 ml of 0,5X TBE is recommended. A maximum of 50 ml of agarose is needed.
  • Can use as little as 30 ml for analytical runs with small sample volumes
  • One or two comb positions for maximum of 40 samples throughput
  • Two precision-fit PVC dams snap snuggly into grooves for leakproof gel casting in gel bed
  • Since gels are cast in place, they are ready to run without casting tray or gel handling
  • Less than 15 minutes run time required for elution of DNA in dialysis bags
Delivery information: Mini Fast Sub complete system includes: UV transparent base with grooves for gel casting dams, 2 comb slots and built in storage compartment, platinum electrodes, 2 gel casting dams, 2 combs, safety cover with thread-on power leads.