Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 1:00 am CET Saturday through 0:30 am Monday. Orders placed will be processed after maintenance is complete.

L(+)-Arginine USP free base

Supplier: VWR Chemicals
Warning
Synonyms: H-L-Arg-OHArginine(S)-(+)-ArginineH-Arg-OH

0953-500GEA 0 INR InStock
0953-500G 0953-100G
L(+)-Arginine USP free base
L(+)-Arginine


Formula: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 238 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1
UN: 0000
Merck Index: 13,00785
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty