Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydroxyurea 97-103%

Supplier: VWR Chemicals



Formula: CH₄N₂O₂
MW: 76,05 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 142 - 144 °C
Storage Temperature: Ambient
MDL Number: MFCD00007943
CAS Number: 127-07-1
EINECS: 204-821-7
UN: 0000
Merck Index: 13,04874
Specification Test Results
Heavy Metals 0.003%
Appearance White to off
white crystalline powder 1 %
Purity (dried basis) 97 % - 103 %
Residue on Ignition 0.5 %
Urea and related compounds Pass

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 5 g 1B1368-5G
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty