3-Chloro-5-(pentafluorothio)benzonitrile ≥97%
|
Danger
|
H33840.MDEA
0
INR
H33840.MD
H33840.03
3-Chloro-5-(pentafluorothio)benzonitrile ≥97%
3-Chloro-5-(pentafluorothio)benzonitrile
Formula:
C₇H₃ClF₅NS MW: 263,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201047 CAS Number: 159727-28-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...