Ovens with forced convection
VWRU414005-119EA 0 INR
VWRU414004-564 VWRU414005-117 VWRU414005-118 VWRU414005-119 VWRU414005-114 VWRU414005-115 VWRU414005-116
Ovens with forced convection
Forced air ovens are ideal for demanding drying and heating processes that require precise temperature accuracy with temperature ranges between 70 to 250 °C. The forced air circulation provides uniform heating, precise temperature control, and fast drying using a blower that circulates heated air in a horizontal airflow pattern. Each unit has excellent temperature recovery time of 6 to 9 minutes depending on size and temperature setting. A PT100 temperature sensor is included for repeatability and stability over a long period. A locking mode assists in preventing unintentional temperature changes.
  • Advanced, adaptive, microprocessor control system with rotary knob
  • resulting in superior temperature accuracy
  • High-quality LCD display with back-light
  • Superior heat insulation provided by heat-resistant rubber packing and Air Flow Layer (AFL)
  • Over temperature & over current protection, sensor error detection
Units are constructed with a durable powder-coated exterior, stainless steel interior, glass wool insulation material, high-temperature grade foamed silicone rubber door gasket, and 1,3 stainless steel ventilation caps with outer exhaust duct of 4 cm.
  • Advanced, adaptive, microprocessor control system with rotary knob
  • resulting in superior temperature accuracy
  • High-quality LCD display with back-light
  • Superior heat insulation provided by heat-resistant rubber packing and Air Flow Layer (AFL)
  • Over temperature & over current protection, sensor error detection
Units are constructed with a durable powder-coated exterior, stainless steel interior, glass wool insulation material, high-temperature grade foamed silicone rubber door gasket, and 1,3 stainless steel ventilation caps with outer exhaust duct of 4 cm.
Delivery information: Supplied with 2 stainless steel shelves.