Glycerine solution 20%, Biotechnology Grade, sterile
Warning
|
Synonyms:
Propane-1,2,3-triol
, Glycerol
, 1,2,3-Propanetriol
E550-100MLEA
6621
INR
E550-100ML
Glycerine solution 20%, Biotechnology Grade, sterile
Glycerine
Used as a cryopreservative in bacterial preservation media.
- Sterile
- Used in cell culture applications
Formula:
HOCH₂CH(OH)CH₂OH MW: 92,09 g/mol |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 |
Specification Test Results
Appearance | Colorless haze |
Free liquid | Pass |
Bioburden Test | None |
Glycerol ID Test | Pass |
Water (KF) | 76 – 84 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...