Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Penicillin : Streptomycin : Amphotericin B , Biotechnology Grade

Supplier: VWR Chemicals

Aseptic, gamma-irradiated, tested for tissue culture. 10000 U/ml penicillin, 10 mg/ml streptomycin and 25 μg/ml amphotericin B when reconstituted in 20 ml of sterile water.


Specification Test Results
25 ug/mL
Penicillin G Sodium Salt 10000 u/mL
Streptomycin sulfate 10 mg/mL

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Vial 20 ml E485-20ML
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty