Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glycogen solution 20 mg/ml for biotechnology

Supplier: VWR Chemicals

Glycogen is an inert carrier that increases the recovery of nucleic acids after alcohol precipitation. Insoluble in ethanol or isoproanol, glycogen traps the nucleic acids during precipitation and forms a visible pellet upon centrifugation. As an inert co-precipitant, glycogen efficiently recovers from dilute solutions DNA or RNA molecules ranging in size from small oligonucleotides to megabase molecules.

Glycogen is a highly purified branched chain carbohydrate that does not add exogenous nucleic acids to the sample as do other coprecipitants such as yeast RNA or tRNA. VWR’s glycogen solution, 20 mg/ml, is free of host DNA and is functionally tested for DNA precipitation. It will not affect DNA/RNA concentration measurements determined by A260/280 ratios. At concentrations less than ~8 μg/μl, glycogen will not interfere with downstream applications including most enzymatic reactions or electrophoresis.


Formula: C₂₄H₄₂O₂₁
MW: 666,579 g/mol
CAS Number: 9005-79-2
Specification Test Results
Bioburden Test NONE
DNA Contamination none
DNase (EQC) none detected
Performance/Gel Visibility Pass
RNase none detected

 

ORDER

Expand All / Collapse All
Pack type Pk Info Pk VWR Catalog Number Quantity
Plastic tube 2×0.5 ml 1 Pack N632-2X0.5ML
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty