2-Chloro-3-fluorobenzonitrile ≥97%

Warning

H33130.03EA 0 INR
H33130.03 H33130.MD
2-Chloro-3-fluorobenzonitrile ≥97%
2-Chloro-3-fluorobenzonitrile
Formula: C₇H₃ClFN
MW: 155,56 g/mol
Melting Pt: 55…56 °C
Storage Temperature: Ambient
MDL Number: MFCD03407966
CAS Number: 874781-08-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR