3-Chloro-5-fluoro-4-methoxy-2-methylbenzaldehyde ≥97%
|
Warning
|
H34338.MDEA
0
INR
H34338.MD
H34338.03
3-Chloro-5-fluoro-4-methoxy-2-methylbenzaldehyde ≥97%
3-Chloro-5-fluoro-4-methoxy-2-methylbenzaldehyde
Formula:
C₉H₈ClFO₂ MW: 202,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201016 CAS Number: 1373920-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...