VWR® Economy Line Chairs and Stools
Supplier: Avantor
132-0500EA 30216 INR
VWRI132-0500 VWRI132-0499
VWR® Economy Line Chairs and Stools
All chairs and stools are available with a star base which is made from high strength sectional steel tubing and is particularly robust and durable. The flat design prevents trip hazards.
  • Anatomically-designed seat and backrest
  • Washable, easy to clean, robust and durable
  • Seat height adjustment by gas-lift
Glides: The glides are wear-proof plastic and have a large surface area and offering a high level of stability.
Castors: Dual-wheel castors with load-sensitive brakes have to be right for the floor. All chairs are supplied with soft castors for hard floors as standard. Hard castors for soft floors are also available as an option.
Stop&Go technology: For safety reasons chairs whose seat is more than 650 mm high must not be movable whilst in use. With innovative Stop&Go castors these chairs are only mobile when no one is sitting on them.
As well as the need for maximum hygiene and easy cleaning, laboratory chairs also have to meet a number of other requirements associated with routine laboratory tasks: Flexibility, in terms of the work applications and they must not take too much space.