Would you like to log in again?
Liquid detergent, low foaming, Det-O-Jet®
Supplier: ALCONOX
Det-O-Jet®
Low foaming, highly alkaline liquid detergent which is ideal for mechanical washers, power-spray systems, and ultrasonic cleaners, including those equipped with automatic feed and proportioning devices. For removing soil, grit, organic contaminants, inorganic residue, chemicals, and solvents from glass, stainless steel, ceramic, cement, and alkali-resistant plastics and metals.
  • Concentrated, free-rinsing formula is highly emulsifying and leaves no interfering residue
  • Completely soluble in hot or cold, hard or soft water - protects and lubricates working parts of washers
  • Typical dilution range 0,5 - 1% depending on application, with pH of 12,4 for 1% solution
  • Biodegradable
  • USDA authorised
ORDER
Expand All / Collapse All
Description Capacity Pk VWR Catalog Number Quantity
Liquid detergent, Det-O-Jet® 0,95 l 0,95 l ALCO1632
Liquid detergent, Det-O-Jet® 3,8 l 3,8 l ALCO1601
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty