Pipette tips, ultra low retention
Supplier: Brand
613-2350EA 0 INR
613-2419 613-2418 613-2434 613-2435 613-2349 613-2423 613-2350 613-2420 613-2345 613-2404 613-2348 613-2406 613-2417 613-2421 613-2347 613-2405 613-2422 613-2346
Pipette tips, ultra low retention
Pipette Tips
The homogeneous, defect-free surface of these tips has an extremely low surface tension – over 50% less than PTFE. This significantly reduces sample loss and provides substantially higher reproducibility when working with critical media. The tips have a high chemical resistance.
  • No leaching out of additives, no siliconisation of the surface
  • High chemical resistance: Ideal when working with solvents
  • All tips up to 1000 µl are free of DNA (< 40 fg), RNase (< 8.6 fg), endotoxins (< 1 pg) and ATP (< 1 fg)
  • Tips can be autoclaved at 121 °C (2 bar) without damaging the material properties
  • BIO-CERT® tips correspond to the highest quality grade: They are sterile, free from human DNA, RNases, endotoxins and ATP
Ideal for working with solvents and biological samples that contain detergents such as Triton® X-100, SDS and Tween.