Would you like to log in again?
Benchtop water activity meter, HygroLab C1
Supplier: ROTRONIC INSTRUMENTS (UK) LTD
ROTRHYGROLABC1EA 0 INR InStock
ROTRHYGROLABC1 710-0996
Benchtop water activity meter, HygroLab C1
Hygrometers Digital Thermo-Hygrometers
The HygroLab C1 is a high-end device for water activity measurements. Versatile and intuitive, the grade indicator software can be used on a PC to provide remote monitoring with charting and data recording functions. It provides an important indicator for the shelf life of food, pharmaceutical and cosmetic products which contribute to the occurrence and growth of microorganisms.
  • Four input channels for HC2 probes
  • Audible alarm to indicate completed measurement
  • Recalibration by the customer with the appropriate standards
  • Quick function for fast measurement results: four to five minutes
  • Measurement of water activity, relative humidity and temperature
  • Saves up to 2000 data records with aw, % RH, °C/°F, date and time
Delivery information: Set includes bench top instrument (HygroLab C1), software (HW4-P-QUICK-V3), measurement probe (HC2-AW), sampler holder (WP-40), disposable sample containers, 100 each (PS-14 and PS-40), humidity standards (5) and USB cable.
* Only for water activity meters i.e. HygroPalm 23-AW-A and HygroLab C1.
Order Now
ORDER
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty