Would you like to log in again?
Radio signal digital thermometer with bottle probe, Traceable®
Supplier: VWR Collection
Traceable®
This radio-signal thermometer can read refrigerator temperatures from up to 100 feet away. They can provide readings from up to three remote refrigerator sensors, to a main unit (frequency of 433 MHz).
  • Distinctive alarm sounds when temperature rises above/falls below high and low set points, set in 1 °C increments
  • Sealed GRAS (Generally Recognised as Safe by FDA) bottle insulated from rapid temperature changes
  • Easy to read two-line display: Data update every 30 seconds, minimum/maximum memory
Certifications: Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Supplied with a main unit, a remote unit, batteries, flip-open stand, wall mount, VELCRO® and Traceable® calibration certificate.
ORDER
Expand All / Collapse All
Description Accuracy Range Resolution W×D×H Weight Pk VWR Catalog Number Quantity
Wireless radio-signal thermometer with remote refrigerator bottle probe module, Traceable® ±1,0 °C −30…+70 °C 0,1 °C 108×114×19 mm 170 g 1 620-1982
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ACCESSORIES
Expand All / Collapse All
Accessories
Description W×D×H Weight Pk VWR Catalog Number Quantity
Remote refrigerator bottle probe module, Traceable® 59×88×19 mm 85 g 1 620-1983
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty