Safety ankle boots, lace-up, Bac'Run® Exono
Supplier: Honeywell Safety Products
Bac'Run®
114-4538EA 0 INR
114-4543 114-4546 114-4540 114-4539 114-4545 114-4536 114-4544 114-4537 114-4541 114-4542 114-4538
Safety ankle boots, lace-up, Bac'Run® Exono
Shoes and Boots
Safety shoes made from water resistant microfibre, featuring flame resistant Nomex® stitching.
  • Metal-free, light and flexible, puncture resistant Flexium™ midsole
  • Fully lined in highly absorbent Poromax® fabric
  • Light, metal-free injected polymer Spacium™ toecap (200 J)
  • Ergonomical, anti-static, antibacterial insole can be removed
  • POLYNIT outsole in PU/nitrile provides high slip and abrasion resistance, excellent heat resistance (300 °C by contact)
Recommended for indoor work, dedicated to heavy or light chemical industries.
Colour: Black
Certifications: EN ISO 20345:2011; S3, HRO, SRC