Would you like to log in again?
E.Z.N.A.® DNA Probe Purification Kit
Supplier: VWR - Omega Bio-Tek
The E.Z.N.A.® DNA Probe Purification Kit allows for the rapid and convenient recovery of up to 20 μg DNA from random-primer or nick labelling reactions. This kit can also be used for general DNA clean up, where as other methods may lead to nuclease contamination or loss of samples. HiBind® DNA spin-columns and optimised buffers facilitate the isolation of DNA free of nucleotides, unincorporated labels, enzymes and salts. Centrifugation and vacuum protocols are both included, and are designed to be simple and fast, while ensuing minimal DNase/RNase contamination.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Pk VWR Catalog Number Quantity
200 Tests D6538-02 Unavailable
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty