Would you like to log in again?
Stock Your Lab With Clearance Savings.Limited-time Discounts on Lab essentials, Equipment and more
E.Z.N.A.® MicroElute DNA Clean-Up Kit
Supplier: OMEGA BIO-TEK
E.Z.N.A.® MicroElute®
D6296-01EA 0 INR InStock
D6296-01 D6296-02
E.Z.N.A.® MicroElute DNA Clean-Up Kit
Nucleic Acid Purification Systems and Kits DNA/RNA Clean-Up Kits
The E.Z.N.A.® MicroElute DNA Clean-Up Kit is designed to purify DNA fragments from enzymatic reactions (such as labelling reactions) with a small elution volume of 10 to 15 μl. This method uses HiBind® DNA columns to recover DNA bands from 100 bp to 10 kb, free of oligonucleotides, nucleotides, and enzymes in yields exceeding 85%. Binding conditions are adjusted by the addition of a specially formulated buffer, and the sample is applied to a specially designed HiBind® DNA MicroElute column. Following a rapid wash step, DNA is eluted with as little as 10 μl deionised water or Elution Buffer ready for downstream applications.
Order Now
ORDER
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty