Would you like to log in again?
E.Z.N.A.® Soil RNA Kit
Supplier: VWR - Omega Bio-Tek
E.Z.N.A.®
The E.Z.N.A.® Soil RNA Kit is designed to isolate high quality of total RNA from soil samples typically containing humic acid and inhibitor of RT-PCR. This kit has been successfully used to isolated RNA from forest, river, glass, rich soil samples. Isolated RNA can be used for most downstream applications including RT-PCR and Northern blotting.
  • Optimised protocol ensure reproducible results
  • Purified RNA suitable for most applications
ORDER
Expand All / Collapse All
Description Sample type Sample size Pk VWR Catalog Number Quantity
E.Z.N.A.® Soil RNA Kit Soil samples 500 mg 50 Tests R6825-01
E.Z.N.A.® Soil RNA Kit Soil samples 500 mg 5 Tests R6825-00
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty