Would you like to log in again?
E.Z.N.A.® Mollusc RNA Kit
Supplier: OMEGA BIO-TEK
E.Z.N.A.®
R6875-00EA 0 INR InStock
R6875-00
E.Z.N.A.® Mollusc RNA Kit
Nucleic Acid Purification Systems and Kits RNA Purification Systems and Kits
The E.Z.N.A.® Mollusc RNA Kit is designed for the efficient isolation of total RNA from mollusc, arthropod, roundworm, flatworm, and other invertebrate tissue samples rich in mucopolysaccharides. Samples are homogenised and lysed in a high salt buffer containing CTAB, and extracted with chloroform to remove mucopolysaccharides and to denature proteins. Following a rapid precipitation step, binding conditions are adjusted and RNA is further purified using HiBind® RNA spin columns. Purified RNA is suitable for downstream applications such as RT-PCR, Poly A+ (mRNA) purification, and hybridisation techniques.
  • RNA isolation in less than 60 minutes
  • Optimised protocol ensure reproducible results
  • Purified RNA suitable for most applications
Order Now
ORDER
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty