E.Z.N.A.® Tissue RNA Kit
Supplier: Omega Bio-Tek
OMEGR6688-01 OMEGR6688-00
E.Z.N.A.® Tissue RNA Kit
E.Z.N.A.® Tissue RNA Kit provides a rapid and easy method for the isolation of total RNA from difficult-to-lyse animal tissue samples including skeletal muscle, heart, and connective tissues. The Tissue RNA protocol integrates a Proteinase K digestion step in the RNA isolation procedure to breakdown the proteins. This step ensures easy and more efficient isolation of total RNA from fibre-rich tissues allowing improved yield and RNA quality. Purified RNA is ready for most downstream applications such as RT-PCR, Northern blotting, Poly(A)+ purification, nuclease protection, and in vitro translation.
  • High quality RNA in under 60 minutes
  • Optimised buffers ensure the reproducible results
  • No organic extractions
  • Purified RNA suitable for most downstream applications