Would you like to log in again?
E.Z.N.A.® HP Viral DNA/RNA Kit
Supplier: VWR - Omega Bio-Tek
The E.Z.N.A.® HP Viral DNA/RNA Kit provides a faster and easier way to purify viral RNA and DNA. By using a proprietary buffer to concentrate viral nucleic acids from plasma, serum, and other body fluids, this kit provides an enhanced sensitivity in viral monitoring and other applications where the higher viral nucleic acid recovery efficiency is required.
  • Rapid isolation of high quality, ready-to-use viral DNA/RNA
  • No phenol/chloroform extractions
  • Complete removal of contaminants and inhibitors
  • Consistent, high quality yields
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Pk Average yield VWR Catalog Number Quantity
E.Z.N.A.® HP Viral DNA/RNA Kit 5 Tests \<10 µg R6873-00 Unavailable
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty