Would you like to log in again?
E-Z 96® Viral RNA Kit
Supplier: Omega Bio-Tek
E-Z 96®
Based on the same principle of E.Z.N.A.® Viral RNA kit, the E-Z 96® Viral RNA Kit is designed for simultaneously isolating viral RNA from up to 96 cell-free samples in 96-well plate format. E-Z 96® Viral RNA Kit is designed for isolation of Viral RNA from cell free fluids such as plasma, serum, urine, and cell culture supernatant. The procedure completely removes contaminants and enzyme inhibitors making viral RNA isolation fast, convenient, and reliable. This kit has been tested for isolating viral nucleic acids from hepatitis A, C, HIV. The kit is also suitable for isolation of total RNA from cultured cells, tissues, and bacteria. RNA purified using the E-Z 96® Viral RNA method is ready for applications such as RT-PCR.
  • No phenol/chloroform extractions
  • RNA is suitable for downstream applications
ORDER
Expand All / Collapse All
Description Sample type Sample size Pk VWR Catalog Number Quantity
E-Z 96® Viral RNA Kit (1×96) Acellular fluids, Cell culture supernatant, Plasma, Serum, Urine 150 µl 1 KIT R1074-00
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty