Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Ethoxy-3'-(trifluoromethyl)acetophenone ≥97%

Supplier: Alfa Aesar
Warning

H34448.03EA 0 INR InStock
H34448.03 H34448.06
4'-Ethoxy-3'-(trifluoromethyl)acetophenone ≥97%
4'-Ethoxy-3'-(trifluoromethyl)acetophenone


Formula: C₁₁H₁₁F₃O₂
MW: 232,20
Storage Temperature: Ambient
MDL Number: MFCD13196590
CAS Number: 851263-13-3
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty