D-(-)-Luciferin
Supplier: PanReac AppliChem
Synonyms:
(S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid
A1029.0050EA
0
INR
A1029.0050
D-(-)-Luciferin
D(-)-Luciferin
Formula:
C₁₁H₈N₂O₃S₂ MW: 280,3228 g/mol |
MDL Number:
MFCD00042929 CAS Number: 2591-17-5 EINECS: 219-981-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...