Would you like to log in again?
E.Z.N.A.® SQ Plant DNA Kit
Supplier: OMEGA BIO-TEK
E.Z.N.A.®
D3095-01EA 0 INR InStock
D3095-01 D3095-00
E.Z.N.A.® SQ Plant DNA Kit
Nucleic Acid Purification Systems and Kits Genomic DNA Purification Systems and Kits
The E.Z.N.A.® SQ Plant DNA Kit provides a reliable method for the isolation of high molecular weight genomic DNA from various plant species and tissues. This kit can also be used to isolate DNA from fungal tissues. The solution-based system can process single or multiple samples simultaneously in less than 60 minutes. The convenient purification procedure removes contaminants and enzyme inhibitors such as proteins and divalent cations. The SQ Plant DNA protocol can be easily scaled up or down to accomodate large or small samples and downstream applications. DNA purified using the SQ Plant DNA Kit is suitable for most downstream applications such as PCR, Southern blotting and restriction enzyme digestion.
  • Isolation of high molecular weight genomic DNA
  • No phenol/chloroform extractions
  • Sample versatility - DNA purification from 0,01 to 30 g plant and fungi samples
  • Process single or multiple samples in less than 90 minutes
  • Large yields of pure DNA
Order Now
ORDER
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty