VWR® Condensers, Allihn
Supplier: Avantor
VWR®
533-0065EA 819 INR
533-0065 533-0064 533-0063
VWR® Condensers, Allihn
Condensers
Borosilicate glass, grade 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Condenser for Soxhlet extraction with glass tubing connector.
Condenser for Soxhlet extraction with glass tubing connector.