VWR® Burettes, PTFE Stopcock
Supplier: Avantor
VWR®
612-4245EA 1221 INR
612-4245 612-4247 612-4248
VWR® Burettes, PTFE Stopcock
Burettes
Borosilicate glass 3,3, class AS.
Burette with screw type needle valve PTFE stopcock. With LOT certificate.
Accuracy as per class AS DIN 12700 ISO 385.