Precision thermometer PT1000, TD 131
620-1933EA 19330 INR
VWRI620-1933
Precision thermometer PT1000, TD 131
This ready to use thermometer, with removeable probe, features automatic or manual choice of the measuring range. It can be used for general purpose applications in air, gas, liquid and solids measurements (depending on the probe type). It is suitable in pharma and biotech companies (production and labs), in the food industry (dairy products, fish, meat and healthcare) blood, government institutes and environment.
  • Low battery warning indicator
  • Calibratable: Offset and slope input
  • Maximum/minimum memory and 'HOLD' function
Delivery information: Delivered in a plastic case with general purpose PT1000 probe PTZ341 (‒70 to +200 °C), with plastic handle and calibration certificate.