Bromophenol blue sodium salt
Synonyms:
Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate
, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt
, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate
, Bromophenol blue water soluble
M116-50GEA
9178
INR
M116-50G
Bromophenol blue sodium salt
Bromophenol blue sodium salt
Bromophenol blue sodium salt is a proteomics grade tracking dye specially suited for nucleic acid and protein gels.
Designed as a yellow to blue pH indicator: 3,0 to 4,6.
Formula:
C₁₉H₉Br₄NaO₅S MW: 691,95 g/mol |
MDL Number:
MFCD00013793 CAS Number: 34725-61-6 |
Specification Test Results
Clarity of Solution | Pass |
Protease | none detected |
Visual Transition Interval (pH 3.0 – 4.6) | Pass |
Appearance | Light red to purple powder. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...