Would you like to log in again?
Laemmli loading buffer, 4X
Supplier: VWR Chemicals
M337-25MLEA 8418 INR InStock
M337-25ML M337-5ML
Laemmli loading buffer, 4X
Buffers Electrophoresis Buffers Loading Buffers
Laemmli loading buffer, 4X is formulated for use in Laemmli SDS-PAGE, a widely used system for protein electrophoresis. Mixing Laemmli loading buffer, 4X with samples reduces disulphide bonds and denatures the proteins due to the 2-mercaptoethanol and SDS detergent content of the buffer. The binding of SDS to denatured protein imparts uniform negative charge density so that separation on the gel is based on size. Glycerol is included in the loading buffer as a density agent to ensure the samples sink into the sample wells, while bromophenol blue serves as tracking dye to monitor electrophoretic progression.
Order Now
ORDER
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty