Would you like to log in again?
Mitochondrial protein isolation buffer
M328-30MLEA 6263 INR InStock
M328-30ML
Mitochondrial protein isolation buffer
Buffers Extraction Buffers
Mitochondrial protein isolation buffer for enrichment of mitochondrial proteins.
  • Entire extraction procedure takes less than an hour
  • Single component
  • Compatible with Western blotting and ELISA assays
Cells are lysed with a syringe and then centrifuged to separate the cytosolic fraction from an enriched mitochondrial fraction. The collected fractions may be used in downstream applications, including Western blotting and ELISA assays.
Order Now
ORDER
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty