Would you like to log in again?
Total Protein Cell Lysis Buffer
M329-10MLEA 2340 INR InStock
M329-10ML
Total Protein Cell Lysis Buffer
Total Protein Cell Lysis Buffer is a nonionic detergent lysis buffer that enables efficient solubilisation of the plasma and intracellular membranes, breaks weak intermolecular bonds, and solubilises most proteins. Total Protein Cell Lysis Buffer reduces protein denaturation, protein complex disruption and loss of enzymatic activity since it contains less detergent than a typical RIPA lysis buffer. Simply, resuspend cells in Total Protein Cell Lysis Buffer and vortex vigourously. Incubate the cell suspension at 4 ºC with shaking for 30 minutes. Vortex vigorously for 30 seconds and pellet cellular debris. The supernatant contains the total protein lysate.
Order Now
ORDER
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty